Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD19 Antibody (1D3), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB224965APC Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB224965APC 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB224965APC Supplier Novus Biologicals Supplier No. NBP224965APC
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD19 Monoclonal specifically detects CD19 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Flow (Cell Surface), Product Image.

Specifications

Antigen CD19
Applications Flow Cytometry
Classification Monoclonal
Clone 1D3
Concentration 1.0 mg/mL
Conjugate APC
Dilution Flow Cytometry 0.25 ug/10^6 cells, Flow (Cell Surface)
Formulation PBS with 0.05% Sodium Azide
Gene Alias B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, MGC12802, T-cell surface antigen Leu-12
Gene Symbols CD19
Host Species Rat
Immunogen Mouse CD19 Transfected Cell Line
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline B Cell Development and Differentiation Markers, Myeloid derived Suppressor Cell
Primary or Secondary Primary
Gene ID (Entrez) 930
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.