Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD19 Antibody (1D3) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB013706 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB013706 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB013706 Supplier Novus Biologicals Supplier No. NBP280633
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD19 Monoclonal specifically detects CD19 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunoprecipitation, In vivo assay, Flow (Cell Surface), CyTOF-ready.

Specifications

Antigen CD19
Applications Flow Cytometry, Immunoprecipitation, CyTOF
Classification Monoclonal
Clone 1D3
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry : 0.25 ug/10^6 cells, Immunoprecipitation reported in scientific literature (PMID 8566073), In vivo assay reported in scientific literature (PMID 8566073), Flow (Cell Surface), CyTOF-ready
Formulation PBS
Gene Alias B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, MGC12802, T-cell surface antigen Leu-12
Gene Symbols CD19
Host Species Rat
Immunogen Mouse CD19 Transfected Cell Line
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, B Cell Development and Differentiation Markers, Cytokine Research, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Signal Transduction, Stem Cell Lines, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 930
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.