Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD19 Antibody (Bu12), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB164360IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB164360IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB164360IR Supplier Novus Biologicals Supplier No. NB10064360IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD19 Monoclonal specifically detects CD19 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation.

Specifications

Antigen CD19
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone Bu12
Conjugate DyLight 755
Dilution Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, MGC12802, T-cell surface antigen Leu-12
Gene Symbols CD19
Host Species Mouse
Immunogen Human EB-4 Burkitt lymphoma cell line.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, B Cell Development and Differentiation Markers, Cytokine Research, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Signal Transduction, Stem Cell Lines, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 930
Test Specificity NB100-64360 recognizes human CD19, a 95kD cell surface glycoprotein, which is expressed on cells of the B cell lineage and follicular dendritic cells but absent on plasma cells. CD19 is an important signal transduction molecule which is involved in the regulation of B lymphocyte development, activation and differentiation.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.