Learn More
Description
Specifications
Specifications
| Antigen | CD19 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | C19/366 |
| Conjugate | Alexa Fluor 700 |
| Dilution | Immunohistochemistry-Paraffin |
| Gene Alias | B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, MGC12802, T-cell surface antigen Leu-12 |
| Gene Symbols | CD19 |
| Host Species | Mouse |
| Immunogen | Recombinant full-length human CD19 protein (Uniprot: P15391) |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
