Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD19 Antibody (CAT-13.1E10), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB154273 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB154273 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB154273 Supplier Novus Biologicals Supplier No. NBP312064AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Flow Cytometry, Immunofluorescence

Specifications

Antigen CD19
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone CAT-13.1E10
Conjugate Alexa Fluor 532
Dilution Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, MGC12802, T-cell surface antigen Leu-12
Host Species Mouse
Immunogen This antibody was raised by immunising mice against CD19.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, B Cell Development and Differentiation Markers, Cytokine Research, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Signal Transduction, Stem Cell Lines, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 930
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.