Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD19 Antibody (CVID3/155), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP261908A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP261908A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP261908A94 Supplier Novus Biologicals Supplier No. NBP261908AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD19 Monoclonal specifically detects CD19 in Human, Monkey samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CD19
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone CVID3/155
Conjugate Alexa Fluor 594
Dilution Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, MGC12802, T-cell surface antigen Leu-12
Gene Symbols CD19
Host Species Mouse
Immunogen Recombinant full-length human CD19 protein (Uniprot: P15391 )
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 930
Test Specificity Recognizes a protein of 95kDa, identified as CD19. CD19 is a transmembrane glycoprotein that contains two extracellular immunoglobulin-like domains. CD19 is present in both benign and malignant B-cells and is considered to be the most reliable surface marker of this lineage over a wide range of maturational stages. In normal lymphoid tissue, CD19 is observed in germinal centers, in mantle zone cells, and in scattered cells of the inter-follicular areas. Anti-CD19 exhibits an overall immunoreactivity pattern similar to those of the antibodies against CD20 and CD22. However, in contrast to CD20, expression of CD19 is continuous throughout B-cell development and through terminal differentiation of B-cells into plasma cells. Anti-CD19 positivity is seen in the vast majority of B-cell neoplasms commonly at a lower intensity than normal B-cell counterparts. Plasma cell neoplasms are nearly always negative, as are T-cell neoplasms.
Target Species Human, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.