Learn More
CD19 Antibody (HD37) - Chimeric - Azide and BSA Free, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | CD19 |
| Applications | Blocking Assay, CyTOF, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | HD37 |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Blocking/Neutralizing 50 - 200 molar excess, Immunofluorescence 1:10 - 1:500, CyTOF-ready |
| Formulation | PBS with 0.02% Proclin 300. with 0.02% Proclin 300 |
| Gene Alias | B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, MGC12802, T-cell surface antigen Leu-12 |
| Gene Symbols | CD19 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.