Learn More
Invitrogen™ CD19 Polyclonal Antibody

Description
Immunogen sequence: NAESYENEDE ELTQPVARTM DFLSPHGSAW DPSREATSLG SQSYEDMRGI LYAAPQLRSI RGQPGPNHEE DADSYENMDN PDGPDPAWGG GGRMGTWSTR.
Specifications
Specifications
| Antigen | CD19 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 2.84 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 |
| Gene | Cd19 |
| Gene Accession No. | P15391, P25918 |
| Gene Alias | AW495831; B4; B-lymphocyte antigen CD19; B-lymphocyte surface antigen B4; Cd19; CD19 antigen; CD19 molecule; CVID3; differentiation antigen CD19; Leu-12; T-cell surface antigen Leu-12 |
| Gene Symbols | Cd19 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.