Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD1a Antibody (CB-T6), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2452420.2MG
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunofluorescence
CD1a Monoclonal antibody specifically detects CD1a in Human, Mouse samples. It is validated for Flow Cytometry, Immunofluorescence.Specifications
CD1a | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence 1 - 2 ug/ml | |
CD1, CD1a antigen, CD1A antigen, a polypeptide, CD1a molecule, cluster of differentiation 1 A, cortical thymocyte antigen CD1A, differentiation antigen CD1-alpha-3, epidermal dendritic cell marker CD1a, FCB6, HTA1, hTa1 thymocyte antigen, R4, T6, T-cell surface antigen T6/Leu-6, T-cell surface glycoprotein CD1a | |
Mouse | |
49 kDa | |
0.2 mg | |
Dendritic Cell Markers, Immunology | |
909 | |
Human, Mouse | |
Purified |
Flow Cytometry, Immunofluorescence | |
CBT6 | |
Unconjugated | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
CD1A | |
Human CD1a | |
Protein A or G purified | |
RUO | |
Primary | |
At least five CD1 genes (CD1a, b, c, d, and e) are identified. CD1 proteins have been demonstrated to restrict T cell response to non-peptide lipid and glycolipid antigens and play a role in non-classical antigen presentation. CD1a is a non-polymorphic MHC Class 1 related cell surface glycoprotein, expressed in association with Beta-2 microglobulin. Anti-CD1a labels Langerhans cell histiocytosis (Histiocytosis X), extranodal histiocytic sarcoma, a subset of T-lymphoblastic lymphoma/leukemia, and interdigitating dendritic cell sarcoma of the lymph node. When combined with antibodies against TTF-1 and CD5, anti-CD1a is useful in distinguishing between pulmonary and thymic neoplasms since CD1a is consistently expressed in thymic lymphocytes in both typical and atypical thymomas, but only focally in 1/6 of thymic carcinomas and not in lymphocytes in pulmonary neoplasms. Anti-CD1a is reported to be a new marker for perivascular epithelial cell tumor (PEComa). | |
Store at 4C. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction