Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD1b Antibody (737249), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB346549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB346549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB346549 Supplier Novus Biologicals Supplier No. FAB7446MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD1b Monoclonal specifically detects CD1b in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen CD1b
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 737249
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD1, CD1A, CD1b antigen, CD1B antigen, b polypeptide, CD1b molecule, cortical thymocyte antigen CD1B, differentiation antigen CD1-alpha-3, MGC125990, MGC125991, R1, T-cell surface glycoprotein CD1b
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CD1b, Ser18-Ser303, Accession # P29016
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 910
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.