Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD1b Antibody (RIV12), Novus Biologicals™

Mouse Monoclonal Antibody
$268.00 - $524.00
Specifications
Antigen | CD1b |
---|---|
Clone | RIV12 |
Dilution | Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml |
Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP232925F
![]() |
Novus Biologicals
NBP2329250.02MG |
0.02 mg |
Each for $268.00
|
|
|||||
NBP232925A
![]() |
Novus Biologicals
NBP2329250.1MG |
0.1 mg |
Each for $524.00
|
|
|||||
NBP232925B
![]() |
Novus Biologicals
NBP2329250.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
CD1b Monoclonal specifically detects CD1b in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
CD1b | |
Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
P29016 | |
910 | |
Human peripheral lymphocytes | |
Primary | |
Store at 4C. | |
49 kDa |
RIV12 | |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Immunology | |
PBS with 0.05% BSA. with 0.05% Sodium Azide | |
CD1, CD1A, CD1b antigen, CD1B antigen, b polypeptide, CD1b molecule, cortical thymocyte antigen CD1B, differentiation antigen CD1-alpha-3, MGC125990, MGC125991, R1, T-cell surface glycoprotein CD1b | |
CD1B | |
IgG1 κ | |
Protein A purified | |
The mouse monoclonal antibody recognizes CD1b, a 44kDa type I glycoprotein associated with beta2-microglobulin. It is expressed on dendritic cells, Langerhans cells, thymocytes, and T acute lymphoblastic leukemia cells. The CD1 multigene family encodes five forms of the CD1 T-cell surface glycoprotein in human, designated CD1A, 1B, 1C, 1D and 1E. CD1, a type 1 membrane protein, has structural similarity to the MHC class I antigen and has been shown to present lipid antigens for recognition by T lymphocytes. Constitutive endocytosis of CD1B molecules and the differential sorting of MHC class II from lysosomes separate peptide- and lipid antigen-presenting molecules during dendritic cell maturation. CD1B is also expressed in interdigitating cells. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title