Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD1b/c Antibody (B-B5), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB362918 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB362918 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB362918 Supplier Novus Biologicals Supplier No. NBP318096MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD1b/c Monoclonal antibody specifically detects CD1b/c in Human samples. It is validated for Flow Cytometry

Specifications

Antigen CD1b/c
Applications Flow Cytometry
Classification Monoclonal
Clone B-B5
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias BDCA1, CD1, CD1A, CD1B antigen, b polypeptide, CD1C antigen, c polypeptide, cortical thymocyte antigen CD1B, cortical thymocyte antigen CD1C, differentiation antigen CD1-alpha-3, R1, R7, T-cell surface glycoprotein CD1b, T-cell surface glycoprotein CD1c
Host Species Mouse
Immunogen Thymus cells and Jurkat cell line. Hybridoma: Myeloma X63/AG.8653 x Balb/c spleen cells
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline CD Markers
Primary or Secondary Primary
Gene ID (Entrez) 910
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.