Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD1c/BDCA-1 Antibody (CD1C/1603), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB378551 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB378551 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB378551 Supplier Novus Biologicals Supplier No. NBP308841JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD1c/BDCA-1 Monoclonal antibody specifically detects CD1c/BDCA-1 in Human samples. It is validated for ELISA

Specifications

Antigen CD1c/BDCA-1
Applications ELISA
Classification Monoclonal
Clone CD1C/1603
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias BDCA1, c polypeptide, CD1, CD1A, CD1c antigen, CD1c molecule, cortical thymocyte antigen CD1C, differentiation antigen CD1-alpha-3, R7, T-cell surface glycoprotein CD1c
Host Species Mouse
Immunogen Recombinant full-length human CD1c/BDCA-1 protein (Uniprot: P29017)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 911
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.