Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD1c/BDCA-1 Antibody (M241), PE, Novus Biologicals™
SDP

Catalog No. NB310415 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310415 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB310415 Supplier Novus Biologicals Supplier No. NB1002645PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD1c/BDCA-1 Monoclonal antibody specifically detects CD1c/BDCA-1 in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Radioimmune Assays

Specifications

Antigen CD1c/BDCA-1
Applications Immunohistochemistry, Immunoprecipitation, Radioimmune Assays (RIA)
Classification Monoclonal
Clone M241
Conjugate PE
Dilution Immunohistochemistry, Immunoprecipitation, Radioimmunoassay
Formulation PBS
Gene Alias BDCA1, c polypeptide, CD1, CD1A, CD1c antigen, CD1c molecule, cortical thymocyte antigen CD1C, differentiation antigen CD1-alpha-3, R7, T-cell surface glycoprotein CD1c
Host Species Mouse
Immunogen MOLT 4 cell line
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 911
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.