Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD1c/BDCA-1 Antibody (SR1634), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32237250UL
Description
CD1c/BDCA-1 Monoclonal antibody specifically detects CD1c/BDCA-1 in Human samples. It is validated for Western BlotSpecifications
CD1c/BDCA-1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
SR1634 | |
Western Blot 1:500-1:2000 | |
BDCA1, c polypeptide, CD1, CD1A, CD1c antigen, CD1c molecule, cortical thymocyte antigen CD1C, differentiation antigen CD1-alpha-3, R7, T-cell surface glycoprotein CD1c | |
A synthesized peptide derived from human CD1c/BDCA-1 (Uniprot #: P29017) | |
50 μg | |
Cancer, Immunology | |
911 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction