Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD1c/BDCA-1 Antibody (SR1634), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | CD1c/BDCA-1 |
---|---|
Clone | SR1634 |
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
CD1c/BDCA-1 Monoclonal antibody specifically detects CD1c/BDCA-1 in Human samples. It is validated for Western BlotSpecifications
CD1c/BDCA-1 | |
Western Blot 1:500-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
BDCA1, c polypeptide, CD1, CD1A, CD1c antigen, CD1c molecule, cortical thymocyte antigen CD1C, differentiation antigen CD1-alpha-3, R7, T-cell surface glycoprotein CD1c | |
A synthesized peptide derived from human CD1c/BDCA-1 (Uniprot #: P29017) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1634 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cancer, Immunology | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
911 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title