Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD1C Monoclonal Antibody (OTI5A8), TrueMAB™, OriGene

Mouse Monoclonal Antibody
$231.00 - $516.29
Specifications
| Antigen | CD1C |
|---|---|
| Clone | OTI5A8 |
| Concentration | 1 mg/mL |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-253-9731
![]()
|
OriGene
TA505398S |
30 μL |
Each for $231.00
|
|
|||||
50-167-5880
![]()
|
OriGene
TA505398 |
100 μL |
Each for $516.29
|
|
|||||
Description
CD1C Monoclonal antibody specifically detects CD1C in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| CD1C | |
| 1 mg/mL | |
| Unconjugated | |
| Mouse | |
| Human | |
| 911 | |
| Primary | |
| CD1C |
| OTI5A8 | |
| Monoclonal | |
| Liquid | |
| RUO | |
| P29017 | |
| IgG1 | |
| -20° C, Avoid Freeze/Thaw Cycles |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title