Learn More
Description
Specifications
Specifications
| Antigen | CD1d |
| Applications | Flow Cytometry, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | 1B1 |
| Conjugate | Alexa Fluor 405 |
| Dilution | Flow Cytometry, Immunohistochemistry |
| Gene Alias | antigen-presenting glycoprotein CD1d, CD1A, CD1d antigen, CD1D antigen, d polypeptide, CD1d molecule, differentiation antigen CD1-alpha-3, HMC class I antigen-like glycoprotein CD1D, MGC34622, R3, R3G1, T-cell surface glycoprotein CD1d, thymocyte antigen CD1D |
| Gene Symbols | CD1D |
| Host Species | Rat |
| Immunogen | The immunogen for this antibody was CD1d. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
