Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD1d Antibody (1B1), Biotin, Novus Biologicals™
SDP

Catalog No. NB344434 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB344434 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB344434 Supplier Novus Biologicals Supplier No. NBP143461B
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD1d Monoclonal antibody specifically detects CD1d in Mouse, Hamster samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation

Specifications

Antigen CD1d
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone 1B1
Conjugate Biotin
Dilution Flow Cytometry, Immunohistochemistry, Immunoprecipitation
Formulation PBS
Gene Alias antigen-presenting glycoprotein CD1d, CD1A, CD1d antigen, CD1D antigen, d polypeptide, CD1d molecule, differentiation antigen CD1-alpha-3, HMC class I antigen-like glycoprotein CD1D, MGC34622, R3, R3G1, T-cell surface glycoprotein CD1d, thymocyte antigen CD1D
Host Species Rat
Immunogen The immunogen for this antibody was CD1d.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, B Cell Development and Differentiation Markers, Cellular Markers, Mitotic Regulators, Myeloid derived Suppressor Cell, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 912
Target Species Mouse, Hamster
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.