Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD1d Antibody (1B1) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP14346101 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP14346101 0.1 mg
NBP1434615 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP14346101 Supplier Novus Biologicals Supplier No. NBP1434610.1MG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD1d Monoclonal specifically detects CD1d in Mouse, Hamster samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation.

Specifications

Antigen CD1d
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone 1B1
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500
Formulation Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide
Gene Alias antigen-presenting glycoprotein CD1d, CD1A, CD1d antigen, CD1D antigen, d polypeptide, CD1d molecule, differentiation antigen CD1-alpha-3, HMC class I antigen-like glycoprotein CD1D, MGC34622, R3, R3G1, T-cell surface glycoprotein CD1d, thymocyte antigen CD1D
Gene Symbols CD1D
Host Species Rat
Immunogen The immunogen for this antibody was CD1d.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline B Cell Development and Differentiation Markers, Myeloid derived Suppressor Cell
Primary or Secondary Primary
Gene ID (Entrez) 912
Target Species Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.