Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD1d Antibody (51.1), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. N143460APC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N143460APC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N143460APC7 Supplier Novus Biologicals Supplier No. NBP143460APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD1d Monoclonal specifically detects CD1d in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CD1d
Applications Flow Cytometry
Classification Monoclonal
Clone 51.1
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias antigen-presenting glycoprotein CD1d, CD1A, CD1d antigen, CD1D antigen, d polypeptide, CD1d molecule, differentiation antigen CD1-alpha-3, HMC class I antigen-like glycoprotein CD1D, MGC34622, R3, R3G1, T-cell surface glycoprotein CD1d, thymocyte antigen CD1D
Gene Symbols CD1D
Host Species Mouse
Immunogen The immunogen for this antibody was CD1d.
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Alzheimers Research, B Cell Development and Differentiation Markers, Cellular Markers, Mitotic Regulators, Myeloid derived Suppressor Cell, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 912.0
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.