Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD1e Antibody (704407) [DyLight 350], Novus Biologicals™
SDP

Catalog No. NB374790 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB374790 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB374790 Supplier Novus Biologicals Supplier No. FAB7330D
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD1e Monoclonal antibody specifically detects CD1e in Human samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOF

Specifications

Antigen CD1e
Applications Flow Cytometry (Intracellular Staining), CyTOF
Classification Monoclonal
Clone 704407
Conjugate DyLight 350
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD1e molecule, e polypeptide, FLJ17609, membrane-associated, thymocyte antigen CD1E
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CD1e, Met1-Tyr304, Accession # P15812
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 913
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.