Learn More
Invitrogen™ CD1e Polyclonal Antibody

Description
Immunogen sequence: ISWEPSPGAG IRAQNICKVL NRYLDIKEIL QSLLGHTCPR FLAGLMEAGE SELKRKVKP Antigen sequence identity: Mouse 42%; Rat 39%.
Specifications
Specifications
| Antigen | CD1e |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | CD1E |
| Gene Accession No. | P15812 |
| Gene Alias | CD1A; CD1e; CD1E antigen, e polypeptide; CD1e molecule; differentiation antigen CD1-alpha-3; hCD1e; leukocyte differentiation antigen; R2; R2G1; sCD1e; T-cell surface glycoprotein CD1e, membrane-associated; T-cell surface glycoprotein CD1e, soluble; thymocyte antigen CD1E |
| Gene Symbols | CD1E |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.