Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD2 Antibody (299808), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB176122 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB176122 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB176122 Supplier Novus Biologicals Supplier No. FAB18562Z
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD2 Monoclonal specifically detects CD2 in Human samples. It is validated for Flow Cytometry, Western Blot.

Specifications

Antigen CD2
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 299808
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias CD2 antigen, CD2 antigen (p50), sheep red blood cell receptor, CD2 molecule, Erythrocyte receptor, FLJ46032, LFA-2, LFA-3 receptor, lymphocyte-function antigen-2, Rosette receptor, SRBC, T11, T-cell surface antigen CD2, T-cell surface antigen T11/Leu-5
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CD2, Lys25-Asp209, Accession # P06729
Purification Method Protein A or G purified from ascites
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Cancer, Hematopoietic Stem Cell Markers, Immunology, Myeloid derived Suppressor Cell, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 914
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.