Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD2 Antibody (RPA-2.10), Biotin, Novus Biologicals™
SDP

Catalog No. NB337244 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB337244 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB337244 Supplier Novus Biologicals Supplier No. NBP225200B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD2 Monoclonal antibody specifically detects CD2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Neutralization

Specifications

Antigen CD2
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Neutralization
Classification Monoclonal
Clone RPA-2.10
Conjugate Biotin
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Block/Neutralize
Formulation PBS
Gene Alias CD2 antigen, CD2 antigen (p50), sheep red blood cell receptor, CD2 molecule, Erythrocyte receptor, FLJ46032, LFA-2, LFA-3 receptor, lymphocyte-function antigen-2, Rosette receptor, SRBC, T11, T-cell surface antigen CD2, T-cell surface antigen T11/Leu-5
Host Species Mouse
Immunogen A recombinant protein was used as the immunogen for the CD2 antibody.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Cancer, Hematopoietic Stem Cell Markers, Immunology, Myeloid derived Suppressor Cell, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 914
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.