Learn More
Description
Specifications
Specifications
| Antigen | CD2 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Blocking Assay |
| Classification | Monoclonal |
| Clone | RPA-2.10 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 0.1-0.25 ug per 1x10^6 cells. Use reported by customer review, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Block/Neutralize |
| Gene Accession No. | NP_001758 |
| Gene Alias | CD2 antigen, CD2 antigen (p50), sheep red blood cell receptor, CD2 molecule, Erythrocyte receptor, FLJ46032, LFA-2, LFA-3 receptor, lymphocyte-function antigen-2, Rosette receptor, SRBC, T11, T-cell surface antigen CD2, T-cell surface antigen T11/Leu-5 |
| Gene Symbols | CD2 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
