Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD2 Antibody (TS1/8), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP237714UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP237714UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP237714UV Supplier Novus Biologicals Supplier No. NBP237714UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD2 Monoclonal specifically detects CD2 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CD2
Applications Flow Cytometry
Classification Monoclonal
Clone TS1/8
Conjugate DyLight 350
Dilution Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CD2 antigen, CD2 antigen (p50), sheep red blood cell receptor, CD2 molecule, Erythrocyte receptor, FLJ46032, LFA-2, LFA-3 receptor, lymphocyte-function antigen-2, Rosette receptor, SRBC, T11, T-cell surface antigen CD2, T-cell surface antigen T11/Leu-5
Gene Symbols CD2
Host Species Mouse
Immunogen Cytotoxic T lymphocytes.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 914
Test Specificity The mouse monoclonal antibody TS1/8 recognizes CD2, a 50 kDa glycoprotein present on the human peripheral blood T lymphocytes and NK cells; also expressed by all thymocytes. HLDA V; WS Code S025
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.