Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD2 Antibody (TS2/18) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222537
Description
CD2 Monoclonal specifically detects CD2 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, Inhibition Activity, CyTOF-ready, Block/Neutralize.Specifications
CD2 | |
Monoclonal | |
1.0 mg/ml | |
Western Blot 0.5 - 2 μg/mL, Flow Cytometry 3 - 5 μg/ 10^6 cells, Immunoprecipitation 1:10 - 1:500, Inhibition Activity 0.5 μg, CyTOF-ready, Neutralization | |
CD2 antigen, CD2 antigen (p50), sheep red blood cell receptor, CD2 molecule, Erythrocyte receptor, FLJ46032, LFA-2, LFA-3 receptor, lymphocyte-function antigen-2, Rosette receptor, SRBC, T11, T-cell surface antigen CD2, T-cell surface antigen T11/Leu-5 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Western Blot, Flow Cytometry, Immunoprecipitation, Inhibition Assays, CyTOF, Neutralization | |
TS2/18 | |
Unconjugated | |
P06729 | |
CD2 | |
Human LFA-2 (CD2) | |
200 μg | |
Adaptive Immunity, Apoptosis, Immunology | |
914 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction