Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD2 Antibody (TS2/18) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10 - $499.50
Specifications
Antigen | CD2 |
---|---|
Clone | TS2/18 |
Concentration | 1.0 mg/ml |
Applications | Western Blot, Flow Cytometry, Immunoprecipitation, Inhibition Assays, CyTOF, Neutralization |
Classification | Monoclonal |
Description
CD2 Monoclonal specifically detects CD2 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, Inhibition Activity, CyTOF-ready, Block/Neutralize.Specifications
CD2 | |
1.0 mg/ml | |
Monoclonal | |
Purified | |
RUO | |
Human, Rat | |
CD2 antigen, CD2 antigen (p50), sheep red blood cell receptor, CD2 molecule, Erythrocyte receptor, FLJ46032, LFA-2, LFA-3 receptor, lymphocyte-function antigen-2, Rosette receptor, SRBC, T11, T-cell surface antigen CD2, T-cell surface antigen T11/Leu-5 | |
CD2 | |
IgG1 | |
Protein A purified |
TS2/18 | |
Western Blot, Flow Cytometry, Immunoprecipitation, Inhibition Assays, CyTOF, Neutralization | |
Unconjugated | |
Mouse | |
Adaptive Immunity, Apoptosis, Immunology | |
P06729 | |
914 | |
Human LFA-2 (CD2) | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title