Learn More
Description
Specifications
Specifications
| Antigen | CD2 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | TS2/18 |
| Conjugate | Biotin |
| Dilution | Western Blot |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Alias | CD2 antigen, CD2 antigen (p50), sheep red blood cell receptor, CD2 molecule, Erythrocyte receptor, FLJ46032, LFA-2, LFA-3 receptor, lymphocyte-function antigen-2, Rosette receptor, SRBC, T11, T-cell surface antigen CD2, T-cell surface antigen T11/Leu-5 |
| Gene Symbols | CD2 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
