Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD20 Antibody (2709D), PE/Cy7, Novus Biologicals™
SDP

Catalog No. NB312573 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB312573 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB312573 Supplier Novus Biologicals Supplier No. FAB42252PECY7
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD20 Monoclonal specifically detects CD20 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CD20
Applications Flow Cytometry
Classification Monoclonal
Clone 2709D
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias B1, B-lymphocyte antigen CD20, B-lymphocyte cell-surface antigen B1, B-lymphocyte surface antigen B1, Bp35MGC3969, CD20 antigen, CD20 receptor, CD20S7, CVID5, LEU-16, Leukocyte surface antigen Leu-16, Membrane-spanning 4-domains subfamily A member 1, membrane-spanning 4-domains, subfamily A, member 1, MS4A2
Host Species Rabbit
Immunogen Human CD20 Peptide. The epitope recognized by rituximab, Binder M, et al. (2006) 108:1975-8. doi: 10.1182/blood-2006-04-014639. Epub 2006 May 16. PMID: 16705086
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer, Cancer Stem Cells, Cell Biology, Cytokine Research, Immunology, Signal Transduction, Stem Cell Markers, Tumor Biomarkers
Primary or Secondary Primary
Gene ID (Entrez) 931
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.