Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD20 Antibody (AISB12), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP143435C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP143435C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP143435C Supplier Novus Biologicals Supplier No. NBP143435C
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD20 Monoclonal specifically detects CD20 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen CD20
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone AISB12
Conjugate DyLight 650
Dilution Western Blot, Flow Cytometry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias B1, B-lymphocyte antigen CD20, B-lymphocyte cell-surface antigen B1, B-lymphocyte surface antigen B1, Bp35MGC3969, CD20 antigen, CD20 receptor, CD20S7, CVID5, LEU-16, Leukocyte surface antigen Leu-16, Membrane-spanning 4-domains subfamily A member 1, membrane-spanning 4-domains, subfamily A, member 1, MS4A2
Gene Symbols MS4A1
Host Species Rat
Immunogen The immunogen for this antibody was CD20.
Quantity 0.1 mL
Research Discipline Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer, Cancer Stem Cells, Cell Biology, Cytokine Research, Immunology, Signal Transduction, Stem Cell Markers, Tumor Biomarkers
Primary or Secondary Primary
Gene ID (Entrez) 931
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.