Learn More
Description
Specifications
Specifications
| Antigen | CD20 |
| Applications | Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), In vitro Assay |
| Classification | Monoclonal |
| Clone | B-Ly1 |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry <= 1 ug/10^6 cells |
| Gene Alias | B1, B-lymphocyte antigen CD20, B-lymphocyte cell-surface antigen B1, B-lymphocyte surface antigen B1, Bp35MGC3969, CD20 antigen, CD20 receptor, CD20S7, CVID5, LEU-16, Leukocyte surface antigen Leu-16, Membrane-spanning 4-domains subfamily A member 1, membrane-spanning 4-domains, subfamily A, member 1, MS4A2 |
| Gene Symbols | MS4A1 |
| Host Species | Mouse |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
