Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD200/OX2 Antibody (002), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB073021 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB073021 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB073021 Supplier Novus Biologicals Supplier No. NBP290396APC
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD200/OX2 Monoclonal specifically detects CD200/OX2 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen CD200/OX2
Applications Flow Cytometry
Classification Monoclonal
Clone 2
Conjugate APC
Dilution Flow Cytometry
Formulation PBS
Gene Alias antigen identified by monoclonal MRC OX-2, CD200 antigenMOX1, CD200 molecule, MOX2MRC, MRC OX-2 antigen, OX-2, OX-2 membrane glycoprotein
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD200/OX2 (NP_001076017.1; Met 1-Gly 232).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 4345
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.