Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD200/OX2 Antibody (333), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB108838 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB108838 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB108838 Supplier Novus Biologicals Supplier No. NBP289696AF532
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD200/OX2 Monoclonal specifically detects CD200/OX2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CD200/OX2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 333
Conjugate Alexa Fluor 532
Formulation 50 mM Sodium Borate
Gene Alias antigen identified by monoclonal MRC OX-2, CD200 antigenMOX1, CD200 molecule, MOX2MRC, MRC OX-2 antigen, OX-2, OX-2 membrane glycoprotein
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human CD200/OX2 (NP_005935.4; Met1-Gly232).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 4345
Test Specificity Has cross-reactivity in ELISA with: Mouse CD200
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.