Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD200/OX2 Antibody (OX-102), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB165603FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB165603FR 0.125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB165603FR Supplier Novus Biologicals Supplier No. NB10065603FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD200/OX2 Monoclonal specifically detects CD200/OX2 in Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen CD200/OX2
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone OX-102
Conjugate DyLight 680
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias antigen identified by monoclonal MRC OX-2, CD200 antigenMOX1, CD200 molecule, MOX2MRC, MRC OX-2 antigen, OX-2, OX-2 membrane glycoprotein
Gene Symbols CD200
Host Species Mouse
Immunogen Membrane fraction of thioglycollate-elicited rat peripheral cells.
Purification Method Protein G purified
Quantity 0.125 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 4345
Test Specificity NB100-65603 recognizes the rat OX2 (CD200) receptor 1. This antigen is a heavily glycosylated 60-100kD cell surface molecule expressed by cells of the myeloid lineage but not by T or B lymphocytes. This antibody has been shown to block the interaction of OX2 receptor 1 with CD200.
Target Species Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.