Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD200/OX2 Antibody (OX-104), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB064860549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB064860549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB064860549 Supplier Novus Biologicals Supplier No. NB10064860JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD200/OX2 Monoclonal specifically detects CD200/OX2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).

Specifications

Antigen CD200/OX2
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OX-104
Conjugate Janelia Fluor 549
Dilution Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias antigen identified by monoclonal MRC OX-2, CD200 antigenMOX1, CD200 molecule, MOX2MRC, MRC OX-2 antigen, OX-2, OX-2 membrane glycoprotein
Gene Symbols CD200
Host Species Mouse
Immunogen Made to Human CD200
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 4345
Test Specificity NB100-64860 recognizes the human CD200 cell surface antigen, also known as OX2. CD200 is expressed by a subset of B lymphocytes, some endothelial cells and by neurons. Recent studies have suggested that the CD200-CD200 ligand system is of importance in the control of macrophage and granulocyte activation.
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.