Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD200/OX2 Antibody (OX-2), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB10065320V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10065320V 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10065320V Supplier Novus Biologicals Supplier No. NB10065320V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD200/OX2 Monoclonal specifically detects CD200/OX2 in Rat samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunomicroscopy, Immunoprecipitation.

Specifications

Antigen CD200/OX2
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunomicroscopy, Immunoprecipitation
Classification Monoclonal
Clone OX-2
Conjugate DyLight 405
Dilution Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias antigen identified by monoclonal MRC OX-2, CD200 antigenMOX1, CD200 molecule, MOX2MRC, MRC OX-2 antigen, OX-2, OX-2 membrane glycoprotein
Gene Symbols CD200
Host Species Mouse
Immunogen Rat thymocyte membrane glycoproteins
Purification Method Protein G purified
Quantity 0.25 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 4345
Test Specificity NB100-65320 recognizes the rat CD200 cell surface antigen, also known as OX-2, a type I membrane glycoprotein, expressed on rat thymocytes, neurons, follicular dendritic cells, endothelium and at low levels on some B cells and smooth muscle cells. Studies have indicated that the CD200-CD200 ligand system is important in the control of macrophage and granulocyte activation and that this antibody blocks OX-2 mediated T cell costimulation. This product is routinely tested by flow cytometry on rat thymocytes.
Target Species Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.