Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD200R1 Antibody (OX-110) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10063562 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.125 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10063562 0.125 mg
NB017550 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10063562 Supplier Novus Biologicals Supplier No. NB10063562

Rat Monoclonal Antibody

CD200R1 Monoclonal specifically detects CD200R1 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen CD200R1
Applications Flow Cytometry
Classification Monoclonal
Clone OX-110
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:25-1:50
Gene Accession No. Q9ES57
Gene Alias CD200 receptor 1, CD200RMOX2Rcell surface glycoprotein CD200 receptor 1, Cell surface glycoprotein OX2 receptor 1, cell surface glycoprotein receptor CD200, CRTR2, HCRTR2, MOX2 receptor, OX2RCD200 cell surface glycoprotein receptor
Gene Symbols CD200R1
Host Species Rat
Immunogen Fusion protein mCD200RCD4d3+4
Purification Method Protein G purified
Quantity 0.125 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 131450
Test Specificity NB100-63562 recognizes mouse CD200R, a cell surface glycoprotein (also known as OX2R), In mouse CD200R is expressed primarily by peripheral blood monocytes and neutrophils but also by other leucocytes including T-lymphocytes and mast cells (1). Studies suggest that CD200-CD200R interaction may be involved in the control of myeloid cellular function.
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.