Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD200R1 Antibody (OX-110) - BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$206.00
Specifications
Antigen | CD200R1 |
---|---|
Clone | OX-110 |
Concentration | 1.0 mg/mL |
Dilution | Flow Cytometry 1:25-1:50 |
Applications | Flow Cytometry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017550
![]() |
Novus Biologicals
NB100635620.025MG |
0.025 mg |
Each for $206.00
|
|
|||||
NB10063562
![]() |
Novus Biologicals
NB10063562 |
0.125 mg | N/A | N/A | N/A | ||||
Description
CD200R1 Monoclonal specifically detects CD200R1 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD200R1 | |
1.0 mg/mL | |
Flow Cytometry | |
Unconjugated | |
Rat | |
Mouse | |
CD200 receptor 1, CD200RMOX2Rcell surface glycoprotein CD200 receptor 1, Cell surface glycoprotein OX2 receptor 1, cell surface glycoprotein receptor CD200, CRTR2, HCRTR2, MOX2 receptor, OX2RCD200 cell surface glycoprotein receptor | |
CD200R1 | |
IgG2a | |
Protein G purified | |
NB100-63562 recognizes mouse CD200R, a cell surface glycoprotein (also known as OX2R), In mouse CD200R is expressed primarily by peripheral blood monocytes and neutrophils but also by other leucocytes including T-lymphocytes and mast cells (1). Studies suggest that CD200-CD200R interaction may be involved in the control of myeloid cellular function. |
OX-110 | |
Flow Cytometry 1:25-1:50 | |
Monoclonal | |
Purified | |
RUO | |
Q9ES57 | |
131450 | |
Fusion protein mCD200RCD4d3+4 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title