Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD200R1 Antibody (OX108) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP25269402 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP25269402 0.2 mg
NBP25269425 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP25269402 Supplier Novus Biologicals Supplier No. NBP2526940.2MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD200 R1 Monoclonal specifically detects CD200 R1 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen CD200 R1
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone OX108
Conjugate Unconjugated
Dilution Flow Cytometry 1:10 - 1:1000, CyTOF-ready
Formulation PBS with 0.02% Proclin 300. with 0.02% Proclin 300
Gene Alias CD200 receptor 1, CD200RMOX2Rcell surface glycoprotein CD200 receptor 1, Cell surface glycoprotein OX2 receptor 1, cell surface glycoprotein receptor CD200, CRTR2, HCRTR2, MOX2 receptor, OX2RCD200 cell surface glycoprotein receptor
Gene Symbols CD200R1
Host Species Mouse
Immunogen Recombinant protein - hCD200RCD4d3+4.
Purification Method Protein A or G purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Cancer, GPCR, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 131450
Test Specificity Recognizes human CD200R, a receptor for the CD200/OX2.
Target Species Human
Content And Storage Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.