Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD200R1 Antibody (OX108) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00 - $584.50
Specifications
Antigen | CD200 R1 |
---|---|
Clone | OX108 |
Dilution | Flow Cytometry 1:10 - 1:1000, CyTOF-ready |
Applications | Flow Cytometry, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP25269425
![]() |
Novus Biologicals
NBP2526940.025MG |
0.025 mg |
Each for $268.00
|
|
|||||
NBP25269402
![]() |
Novus Biologicals
NBP2526940.2MG |
0.2 mg |
Each for $584.50
|
|
|||||
Description
CD200R1 Monoclonal specifically detects CD200R1 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
CD200 R1 | |
Flow Cytometry 1:10 - 1:1000, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD200 receptor 1, CD200RMOX2Rcell surface glycoprotein CD200 receptor 1, Cell surface glycoprotein OX2 receptor 1, cell surface glycoprotein receptor CD200, CRTR2, HCRTR2, MOX2 receptor, OX2RCD200 cell surface glycoprotein receptor | |
CD200R1 | |
IgG1 | |
Protein A or G purified | |
Recognizes human CD200R, a receptor for the CD200/OX2. |
OX108 | |
Flow Cytometry, CyTOF | |
Unconjugated | |
Mouse | |
Cancer, GPCR, Neuronal Cell Markers, Neuroscience | |
PBS with 0.02% Proclin 300. with 0.02% Proclin 300 | |
131450 | |
Recombinant protein - hCD200RCD4d3+4. | |
Primary | |
Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title