Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD200R1L Antibody (2692A), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB175817 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB175817 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB175817 Supplier Novus Biologicals Supplier No. FAB10198D
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CD200R1L Monoclonal specifically detects CD200R1L in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry.

Specifications

Antigen CD200R1L
Applications Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone 2692A
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias cell surface glycoprotein CD200 receptor 2, CD200 receptor 1-like, CD200R2, CD200RLa
Host Species Rabbit
Immunogen Mouse myeloma cell line NS0-derived mouse CD200R1L, Ile25-Thr220, Accession # Q6XJV6.1
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 344807
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.