Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD200R1L Antibody (6D6C1), PE, Novus Biologicals™
SDP

Catalog No. NB352122 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB352122 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB352122 Supplier Novus Biologicals Supplier No. NBP306388PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD200R1L Monoclonal antibody specifically detects CD200R1L in Human samples. It is validated for ELISA, ELISA

Specifications

Antigen CD200R1L
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 6D6C1
Conjugate PE
Dilution ELISA, Sandwich ELISA Detection
Formulation PBS
Gene Alias cell surface glycoprotein CD200 receptor 2, CD200 receptor 1-like, CD200R2, CD200RLa
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD200R1L (Accession#: AAT00538.1; Met1-Leu239).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 344807
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.