Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD200R1L Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB180016 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB180016 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB180016 Supplier Novus Biologicals Supplier No. NBP306398R
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CD200R1L Polyclonal antibody specifically detects CD200R1L in Human samples. It is validated for ELISA, Immunofluorescence

Specifications

Antigen CD200R1L
Applications ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias cell surface glycoprotein CD200 receptor 2, CD200 receptor 1-like, CD200R2, CD200RLa
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human CD200R1L (AAT00538.1; Met1-Leu239).
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 344807
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.