Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD200R4 Antibody (4), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB316381 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB316381 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB316381 Supplier Novus Biologicals Supplier No. NBP290661MFV500
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD200R4 Monoclonal antibody specifically detects CD200R4 in Mouse samples. It is validated for ELISA

Specifications

Antigen CD200R4
Applications ELISA
Classification Monoclonal
Clone 4
Conjugate mFluor Violet 500 SE
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias F630107N04Rik, MCD200, MCD200RLa
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD200R4 (Accession#: NP_997127.1; Met 1-Thr 241).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 239849
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.