Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD200R4 Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB132166 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB132166 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB132166 Supplier Novus Biologicals Supplier No. NBP306093R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CD200R4 Polyclonal specifically detects CD200R4 in Mouse samples. It is validated for ELISA.

Specifications

Antigen CD200R4
Applications ELISA
Classification Polyclonal
Conjugate DyLight 550
Dilution ELISA
Formulation 50mM Sodium Borate
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Mouse CD200R4 (NP_997127.1; Met 1-Thr 241).
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 239849
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.