Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD201 (EPCR) Monoclonal Antibody (RCR-227), APC, eBioscience™
GREENER_CHOICE

Catalog No. p-7169177
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-112-3100 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-112-3100 Supplier Invitrogen™ Supplier No. 17201842
Only null left
Add to Cart
Edge
Add to Cart

Rat Monoclonal Antibody

Description: The monoclonal antibody RCR-227 reacts with human CD201, which is also known as the endothelial protein C receptor (EPCR), a 46-kDa transmembrane protein related to class I MHC. CD201 was originally identified on vascular endothelium cells, predominantly larger blood vessels. It is also expressed on monocytes, lymphocytes, hematopoietic stem cells, neutrophils, and eosinophils. CD201 serves as a binding site for protein C, activated protein C (APC), and factor VII/VIIa. A soluble form has also been identified in the plasma. CD201 plays a role in controlling coagulation as disruption of its gene in mice caused placental thrombosis and early embryonic death. This cell surface molecule is also involved in the anti-inflammatory response. APC binding to CD201 enables PAR-1 activation that confers cytoprotective effects. In monocytes, APC binding decreases secretion of proinflammatory cytokines, chemokines, and growth factors. Finally, CD201 has been shown to inhibit lymphocyte migration. Applications Reported: This RCR-227 antibody has been reported for use in flow cytometric analysis. Applications Tested: This RCR-227 antibody has been pre-titrated and tested by flow cytometric analysis of HUVECs. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-...

The protein encoded by this gene is a receptor for activated protein C, a serine protease activated by and involved in the blood coagulation pathway. The encoded protein is an N-glycosylated type I membrane protein that enhances the activation of protein C. Mutations in this gene have been associated with venous thromboembolism and myocardial infarction, as well as with late fetal loss during pregnancy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD201 (EPCR)
Applications Flow Cytometry
Classification Monoclonal
Clone RCR-227
Concentration 5 μL/Test
Conjugate APC
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene Procr
Gene Accession No. Q9UNN8
Gene Alias Activated protein C receptor; AI325044; APC receptor; bA42O4.2; CCCA; CCD41; CD antigen CD201; CD201; CD201 antigen; cell cycle, centrosome-associated protein; centrocyclin; Centrosomal protein CCD41; endothelial cell protein C receptor; Endothelial protein C receptor; Epcr; PROCR; protein C receptor; protein C receptor, endothelial
Gene Symbols Procr
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10544
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.