Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD207 (Langerin) Monoclonal Antibody (eBioL31), Biotin, eBioscience™
GREENER_CHOICE

Catalog No. 5011821
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-118-21 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-118-21 Supplier Invitrogen™ Supplier No. 13207582
Only null left
Add to Cart
Edge
Add to Cart

Rat Monoclonal Antibody

Description: The eBioL31 monoclonal antibody reacts with mouse CD207, also known as Langerin, which is expressed in a distinct subset of dendritic cells called Langerhans cells (LC). mouse CD207 is a 48 kDa C-type lectin transmembrane protein that likely plays a role in antigen recognition and uptake. LC are located in the epidermis and upon activation, reduce CD207 expression and begin migrating through the dermis towards lymphatic vessels. Expression of mouse CD207 in LC has been correlated to the presence of Birbeck Granules. Furthermore, the formation of Birbeck Granules has been observed upon transfection of CD207 cDNA into fibroblast cell lines. Western Blotting with eBioL31 reveals a 48kDa band in lysate from mouse ear epidermis, as well as in lysates from cells transfected with mouse CD207 cDNA. Preliminary data from mouse CD207 cDNA transfected cells suggests that eBioL31 can be used for intracellular flow cytometric analysis in combination with the Foxp3/Transcription Factor Staining Bufer Set (Product # 00-5523). The epitope recognized by eBioL31 is in the extracellular domain within the CRD (carbohydrate recognition domain) and therefore can be used to detect both surface and intracellular langerin. Differences in expression has been observed between mouse strains. Applications Reported: This eBioL31 antibody has been reported for use in intracellular staining followed by flow cytometric analysis, and immunohistology staining of frozen tissue sections. Applicat...

Langerin (CD207) is a type II membrane-associated C-type lectin known to be expressed exclusively by Langerhans cells. Lamgerin recognizes mannose residues via its single carbohydrate recognition domain (CRD). Langerin is localized not only on the cell surface, but also intracellularly in close association with Birbeck granules. Transfection of Langerin cDNA into fibroblasts creates a compact network of membrane structures with typical features of Birbeck granules (BG). Langerin is thus a potent inducer of membrane superimposition and zippering leading to BG formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD207 (Langerin)
Applications Flow Cytometry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone eBioL31
Concentration 0.5 mg/mL
Conjugate Biotin
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene CD207
Gene Accession No. Q8VBX4
Gene Alias CD 207 antigen; CD207; CD207 antigen; CD207 antigen, langerin; CD207 molecule; CD207 molecule, langerin; CLEC4K; C-type lectin domain family 4 member K; C-type lectin domain family 4, member K; Langerhans cell specific c-type lectin; langerin; RGD1565913
Gene Symbols CD207
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246278
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.