Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD21 Antibody (1F8), Novus Biologicals™
SDP

Catalog No. NB018457 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018457 0.1 mL
NBP122527 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018457 Supplier Novus Biologicals Supplier No. NBP1225270.1ML
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD21 Monoclonal specifically detects CD21 in Human, Monkey samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CD21
Applications Immunohistochemistry (Paraffin), Immunohistochemistry
Classification Monoclonal
Clone 1F8
Conjugate Unconjugated
Dilution Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation Tissue culture supernatant, containing less than 0.1% sodium azide with Sodium Azide
Gene Accession No. P20023
Gene Alias C3DRSLEB9, CD21, CD21 antigen, Complement C3d receptor, complement component (3d/Epstein Barr virus) receptor 2, complement receptor type 2, Cr2, EBV receptor, Epstein-Barr virus receptor
Gene Symbols CR2
Host Species Mouse
Immunogen Purified receptor for the C3d fragment of the third complement component CR2, from human tonsil.
Purification Method Tissue culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Dendritic Cell Markers, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 1380
Test Specificity This reacts with CD21 antigen of MW 145 kD, present on the surface of mature B cells. The antigen is present at high density on follicular dendritic cells. CD21 is involved in transmitting growth-promoting signals to the interior of the B cell.
Target Species Human
Content And Storage Store at 4°C.
Form Supernatant
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.